siRNA Details
| Gene Symbol: Ccm2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC020535.1 | - | 10 | 2147 | 4240594 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccm2l| Gene Symbol: Ccm2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC020535.1 | - | 10 | 2147 | 4240594 | |