siRNA Details
| Gene Symbol: Yipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205156.1 | NP_001192085.1 | 5 | 493 | ||
| NM_145550.3 | NP_663525.1 | 6 | 574 | |||
| GenBank mRNA | AK081205.1 | - | 6 | 561 | ||
| AK157536.1 | - | 6 | 561 | |||
| AK212572.1 | - | 1 | 40 | |||
| AK214559.1 | - | 1 | 43 | |||
| BC009080.1 | - | 6 | 506 | 2655774 | ||

Yipf1
Genomic Map