siRNA Details
| Gene Symbol: Eva1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145570.2 | NP_663545.2 | 3 | 822 | ||
| GenBank mRNA | BC014699.1 | - | 3 | 755 | 4221746 | |
| BC027150.1 | - | 3 | 777 | 4950341 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Eva1a
Eva1a| Gene Symbol: Eva1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145570.2 | NP_663545.2 | 3 | 822 | ||
| GenBank mRNA | BC014699.1 | - | 3 | 755 | 4221746 | |
| BC027150.1 | - | 3 | 777 | 4950341 | ||
 Back To Top
 Back To Top