siRNA Details
| Gene Symbol: Gys2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145572.2 | NP_663547.2 | 2 | 567 | ||
| GenBank mRNA | AK138992.1 | - | 2 | 275 | ||
| AK149391.1 | - | 2 | 291 | |||
| AW106180.1 | - | 2 | 570 | |||
| BC021322.1 | - | 2 | 428 | 5051685 | ||
| BC158081.1 | - | 2 | 428 | 9007373 | ||

Gys2
Genomic Map