siRNA Details
| Gene Symbol: CCIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005893.2 | NP_005884.2 | 1 | 398 | ||
| GenBank mRNA | AF333334.1 | - | 1 | 396 | ||
| AK313763.1 | - | 1 | 403 | |||
| BC019251.2 | - | 1 | 366 | 4825221 | ||
| Z46967.1 | - | 1 | 278 | |||

CCIN
Genomic Map