siRNA Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145828.3 | NP_665827.2 | 5 | 1114 | ||
| GenBank mRNA | AJ291751.2 | - | 5 | 1031 | ||
| AK083482.1 | - | 5 | 1083 | |||
| AK086804.1 | - | 5 | 1046 | |||
| AK150521.1 | - | 5 | 1067 | |||
| AK153332.1 | - | 5 | 1067 | |||
| AK158056.1 | - | 5 | 1114 | |||
| BC034082.1 | - | 4 | 633 | 4239646 | ||

Xylt2
Genomic Map