siRNA Details
| Gene Symbol: Dmrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145831.3 | NP_665830.1 | 3, 4 | 1009 | ||
| GenBank mRNA | AF539811.1 | - | 3, 4 | 1009 | ||
| AK044891.1 | - | 3, 4 | 1010 | |||
| AK140337.1 | - | 3, 4 | 1001 | |||
| AK140347.1 | - | 3, 4 | 1004 | |||

Dmrt2
Genomic Map