siRNA Details
| Gene Symbol: Cc2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145970.1 | NP_666082.2 | 14 | 1570 | ||
| GenBank mRNA | BC016188.1 | - | 14 | 1570 | 4482793 | |
| BC027028.1 | - | 13 | 1385 | 5363547 | ||
| DQ329239.1 | - | 7 | 620 | |||

Cc2d1a
Genomic Map