siRNA Details
| Gene Symbol: BC027231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145972.4 | NP_666084.1 | 8 | 1053 | ||
| GenBank mRNA | AK144612.1 | - | 8 | 1034 | ||
| AK162291.1 | - | 7 | 895 | |||
| BC027231.1 | - | 8 | 1056 | 3585175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC027231| Gene Symbol: BC027231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145972.4 | NP_666084.1 | 8 | 1053 | ||
| GenBank mRNA | AK144612.1 | - | 8 | 1034 | ||
| AK162291.1 | - | 7 | 895 | |||
| BC027231.1 | - | 8 | 1056 | 3585175 | ||