siRNA Details
| Gene Symbol: Pdlim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253736.1 | NP_001240665.1 | 10 | 1347 | ||
| NM_145978.2 | NP_666090.1 | 10 | 1274 | |||
| GenBank mRNA | BC024556.1 | - | 10 | 1274 | 4990983 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdlim2| Gene Symbol: Pdlim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253736.1 | NP_001240665.1 | 10 | 1347 | ||
| NM_145978.2 | NP_666090.1 | 10 | 1274 | |||
| GenBank mRNA | BC024556.1 | - | 10 | 1274 | 4990983 | |