siRNA Details
| Gene Symbol: Larp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040399.1 | NP_001035489.1 | 5 | 435 | ||
| GenBank mRNA | AK076732.1 | - | 5 | 403 | ||
| AK160155.1 | - | 5 | 435 | |||
| BC025528.1 | - | 3 | 358 | 5324328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Larp1b| Gene Symbol: Larp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040399.1 | NP_001035489.1 | 5 | 435 | ||
| GenBank mRNA | AK076732.1 | - | 5 | 403 | ||
| AK160155.1 | - | 5 | 435 | |||
| BC025528.1 | - | 3 | 358 | 5324328 | ||