siRNA Details
| Gene Symbol: Hip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146001.2 | NP_666113.2 | 11, 12 | 1091 | ||
| GenBank mRNA | AK087586.1 | - | 11, 12 | 1089 | ||
| AK147246.1 | - | 11, 12 | 1047 | |||
| AK147269.1 | - | 11, 12 | 1047 | |||
| AK166346.1 | - | 11, 12 | 1060 | |||
| BC017516.1 | - | 11, 12 | 1084 | 4504441 | ||

Hip1
Genomic Map