siRNA Details
| Gene Symbol: Hip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146001.2 | NP_666113.2 | 31 | 3381 | ||
| GenBank mRNA | AK147246.1 | - | 31 | 3336 | ||
| AK147269.1 | - | 31 | 3336 | |||
| BC017516.1 | - | 31 | 3374 | 4504441 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hip1| Gene Symbol: Hip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146001.2 | NP_666113.2 | 31 | 3381 | ||
| GenBank mRNA | AK147246.1 | - | 31 | 3336 | ||
| AK147269.1 | - | 31 | 3336 | |||
| BC017516.1 | - | 31 | 3374 | 4504441 | ||