siRNA Details
| Gene Symbol: Flcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271356.1 | NP_001258285.1 | 6 | 1119 | ||
| NM_001271357.1 | NP_001258286.1 | 6 | 862 | |||
| NM_146018.2 | NP_666130.1 | 6 | 1159 | |||
| GenBank mRNA | AK039106.1 | - | 6 | 946 | ||
| AK080933.1 | - | 6 | 944 | |||
| AK136071.1 | - | 6 | 850 | |||
| AK148216.1 | - | 6 | 913 | |||
| AK154040.1 | - | 6 | 1141 | |||
| AK161460.1 | - | 6 | 891 | |||
| AK172245.1 | - | 6 | 918 | |||
| AK187898.1 | - | 2 | 57 | |||
| BC025820.1 | - | 6 | 893 | 5099493 | ||

Flcn
Genomic Map