siRNA Details
| Gene Symbol: Gm21975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146023.4 | NP_666135.1 | 4 | 2944 | ||
| Gene Symbol: Evi2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077496.1 | NP_001070964.1 | 2 | 2509 | ||

Gm21975
Genomic Map