siRNA Details
| Gene Symbol: Plekhh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146030.2 | NP_666142.1 | 6 | 1137 | ||
| GenBank mRNA | AK154510.1 | - | 6 | 1153 | ||
| AK164128.1 | - | 6 | 1154 | |||
| BC020025.1 | - | 6 | 1137 | 3490992 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhh3| Gene Symbol: Plekhh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146030.2 | NP_666142.1 | 6 | 1137 | ||
| GenBank mRNA | AK154510.1 | - | 6 | 1153 | ||
| AK164128.1 | - | 6 | 1154 | |||
| BC020025.1 | - | 6 | 1137 | 3490992 | ||