siRNA Details
| Gene Symbol: Plekhh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146030.2 | NP_666142.1 | 13 | 2886 | ||
| GenBank mRNA | AK052371.1 | - | 5 | 1061 | ||
| AK154510.1 | - | 13 | 2893 | |||
| AK164128.1 | - | 13 | 2896 | |||
| BC020025.1 | - | 13 | 2886 | 3490992 | ||
| BC024538.1 | - | 6 | 1289 | |||

Plekhh3
Genomic Map