siRNA Details
| Gene Symbol: Ccdc166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163518.1 | NP_001156990.1 | 1 | 1052 | ||
| NM_146059.2 | NP_666171.2 | 2 | 939 | |||
| GenBank mRNA | AK019123.1 | - | 1 | 749 | ||
| AK137409.1 | - | 1 | 753 | |||
| AK171003.1 | - | 1 | 723 | |||
| BC024536.1 | - | 1 | 660 | 4988004 | ||
| BC047213.1 | - | 1 | 1053 | 6395374 | ||

Ccdc166
Genomic Map