siRNA Details
| Gene Symbol: Krt79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146063.1 | NP_666175.1 | 9 | 1827 | ||
| GenBank mRNA | AK076407.1 | - | 5 | 883 | ||
| BC019155.1 | - | 9 | 1766 | 5065687 | ||
| BC031593.1 | - | 9 | 1827 | 4935395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt79| Gene Symbol: Krt79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146063.1 | NP_666175.1 | 9 | 1827 | ||
| GenBank mRNA | AK076407.1 | - | 5 | 883 | ||
| BC019155.1 | - | 9 | 1766 | 5065687 | ||
| BC031593.1 | - | 9 | 1827 | 4935395 | ||