siRNA Details
| Gene Symbol: Cpped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146067.3 | NP_666179.2 | 2 | 208 | ||
| GenBank mRNA | AK029379.1 | - | 2 | 132 | ||
| AK029442.1 | - | 2 | 231 | |||
| AK032288.1 | - | 2 | 171 | |||
| AK041329.1 | - | 2 | 181 | |||
| AK049721.1 | - | 2 | 209 | |||
| AK133977.1 | - | 2 | 201 | |||
| AK134718.1 | - | 2 | 209 | |||
| BC025476.1 | - | 2 | 129 | |||
| BC034347.1 | - | 2 | 187 | 2654874 | ||

Cpped1
Genomic Map