siRNA Details
| Gene Symbol: Cpped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146067.3 | NP_666179.2 | 4 | 2016 | ||
| GenBank mRNA | AK029379.1 | - | 4 | 1941 | ||
| AK032288.1 | - | 4 | 1979 | |||
| AK049721.1 | - | 4 | 2017 | |||
| AK080876.1 | - | 4 | 1974 | |||
| AK133977.1 | - | 4 | 2010 | |||
| BC025476.1 | - | 4 | 1937 | |||
| BC034347.1 | - | 4 | 1658 | 2654874 | ||

Cpped1
Genomic Map