siRNA Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146068.4 | NP_666180.2 | 4 | 468 | ||
| GenBank mRNA | AK044836.1 | - | 4 | 451 | ||
| AK052832.1 | - | 4 | 433 | |||
| AK075871.1 | - | 4 | 433 | |||
| AK146053.1 | - | 4 | 468 | |||
| AK172902.1 | - | 4 | 421 | |||
| BC089026.1 | - | 4 | 405 | 30615298 | ||

Spidr
Genomic Map