siRNA Details
| Gene Symbol: Zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146073.3 | NP_666185.3 | 3 | 1337 | ||
| GenBank mRNA | AF542387.1 | - | 4 | 1006 | ||
| AK041496.1 | - | 3 | 1020 | |||
| AK041648.1 | - | 4 | 1333 | |||
| AK046719.1 | - | 3 | 1325 | |||
| AK080845.1 | - | 3 | 1341 | |||
| BC059814.1 | - | 3 | 1380 | 6813525 | ||
| BC086758.1 | - | 4 | 393 | |||

Zdhhc14
Genomic Map