siRNA Details
| Gene Symbol: Lemd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146075.2 | NP_666187.2 | 3 | 874 | ||
| GenBank mRNA | AK082131.1 | - | 3 | 681 | ||
| AK143224.1 | - | 3 | 859 | |||
| AY640307.1 | - | 3 | 854 | |||
| BC026588.1 | - | 3 | 184 | 4972900 | ||
| BC050010.1 | - | 1 | 1086 | |||

Lemd2
Genomic Map