siRNA Details
| Gene Symbol: Slc26a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290320.1 | NP_001277249.1 | 9, 10 | 1460 | ||
| GenBank mRNA | BC027076.1 | - | 9, 10 | 1460 | 5361175 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc26a8| Gene Symbol: Slc26a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290320.1 | NP_001277249.1 | 9, 10 | 1460 | ||
| GenBank mRNA | BC027076.1 | - | 9, 10 | 1460 | 5361175 | |