siRNA Details
| Gene Symbol: Zadh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146090.5 | NP_666202.2 | 2 | 1516 | ||
| GenBank mRNA | AK047559.1 | - | 2 | 1520 | ||
| AK081988.1 | - | 2 | 1517 | |||
| AK083082.1 | - | 2 | 1560 | |||
| BC025897.1 | - | 1 | 759 | |||

Zadh2
Genomic Map