siRNA Details
| Gene Symbol: Cbwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146097.3 | NP_666209.1 | 4 | 442 | ||
| GenBank mRNA | AK132291.1 | - | 4 | 428 | ||
| BC018472.1 | - | 4 | 384 | 3661842 | ||
| BC053745.1 | - | 4 | 477 | 6505520 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbwd1| Gene Symbol: Cbwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146097.3 | NP_666209.1 | 4 | 442 | ||
| GenBank mRNA | AK132291.1 | - | 4 | 428 | ||
| BC018472.1 | - | 4 | 384 | 3661842 | ||
| BC053745.1 | - | 4 | 477 | 6505520 | ||