siRNA Details
| Gene Symbol: Afap1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177796.1 | NP_001171267.1 | 20 | 3445 | ||
| NM_001177797.1 | NP_001171268.1 | 19 | 3391 | |||
| NM_146102.2 | NP_666214.1 | 21 | 3497 | |||
| GenBank mRNA | AK048649.1 | - | 21 | 3151 | ||
| AK087449.1 | - | 20 | 3082 | |||
| AK220430.1 | - | 19 | 3014 | |||
| BC031515.1 | - | 20 | 3047 | 3711746 | ||

Afap1l2
Genomic Map