siRNA Details
| Gene Symbol: Aph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146104.3 | NP_666216.1 | 6 | 2214 | ||
| NM_146134.2 | NP_666246.1 | 7 | 1881 | |||
| GenBank mRNA | AK050828.1 | - | 4 | 2438 | ||
| AK078343.1 | - | 4 | 1679 | |||
| AK148400.1 | - | 6 | 2167 | |||
| AK204454.1 | - | 1 | 66 | |||
| BC012406.1 | - | 6 | 2139 | 3483214 | ||

Aph1a
Genomic Map