siRNA Details
| Gene Symbol: Tubb4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146116.2 | NP_666228.1 | 4 | 1101 | ||
| GenBank mRNA | AK002568.1 | - | 1 | 551 | ||
| AK006924.1 | - | 3 | 865 | |||
| AK015901.1 | - | 4 | 1051 | |||
| AK076895.1 | - | 4 | 1058 | |||
| AK146587.1 | - | 4 | 1057 | |||
| AK167022.1 | - | 4 | 1056 | |||
| AK195017.1 | - | 1 | 360 | |||
| BC005547.1 | - | 4 | 1033 | 3585500 | ||
| BC022919.1 | - | 4 | 1040 | 4221189 | ||
| BC083319.1 | - | 4 | 1043 | 5699674 | ||
| Gene Symbol: Zfp646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC098458.1 | - | 2824 | |||

Tubb4b
Genomic Map