siRNA Details
| Gene Symbol: Zfp646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC098458.1 | - | 3201 | |||
| Gene Symbol: Tubb4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146116.2 | NP_666228.1 | 4 | 1478 | ||
| GenBank mRNA | AK002568.1 | - | 1 | 928 | ||
| AK015901.1 | - | 4 | 1428 | |||
| AK076895.1 | - | 4 | 1435 | |||
| AK146587.1 | - | 4 | 1434 | |||
| AK167022.1 | - | 4 | 1433 | |||
| AK214079.1 | - | 1 | 267 | |||
| BC005547.1 | - | 4 | 1410 | 3585500 | ||
| BC022919.1 | - | 4 | 1417 | 4221189 | ||
| BC083319.1 | - | 4 | 1420 | 5699674 | ||
| CJ309661.1 | - | 1 | 308 | |||

Zfp646
Genomic Map