siRNA Details
| Gene Symbol: Dennd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146122.3 | NP_666234.3 | 5 | 502 | ||
| GenBank mRNA | AK137530.1 | - | 4 | 430 | ||
| BC027786.1 | - | 5 | 465 | 5346095 | ||
| BQ444119.1 | - | 5 | 340 | |||
| DQ448594.1 | - | 5 | 247 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191747, XM_008761757, XM_008761756.

Dennd1a
Genomic Map