siRNA Details
| Gene Symbol: Emc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039200.2 | NP_001034289.1 | 6 | 628 | ||
| NM_146157.4 | NP_666269.2 | 6 | 628 | |||
| GenBank mRNA | AK049061.1 | - | 6 | 619 | ||
| AK077496.1 | - | 6 | 618 | |||
| AK089289.1 | - | 6 | 607 | |||
| AK129052.1 | - | 6 | 581 | |||
| AK138637.1 | - | 6 | 600 | |||
| AK161677.1 | - | 6 | 591 | |||
| AK170739.1 | - | 6 | 607 | |||
| BC056952.1 | - | 1 | 76 | |||
| BC129804.1 | - | 6 | 615 | 40087024 | ||
| BC139366.1 | - | 6 | 614 | 8862388 | ||

Emc1
Genomic Map