siRNA Details
| Gene Symbol: Emc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039200.2 | NP_001034289.1 | 23 | 3658 | ||
| NM_146157.4 | NP_666269.2 | 24 | 3667 | |||
| GenBank mRNA | AK049061.1 | - | 24 | 3660 | ||
| AK077496.1 | - | 24 | 3658 | |||
| AK129052.1 | - | 24 | 3624 | |||
| BC129804.1 | - | 24 | 3658 | 40087024 | ||

Emc1
Genomic Map