siRNA Details
| Gene Symbol: Haus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146159.1 | NP_666271.1 | 2 | 527 | ||
| GenBank mRNA | AK049414.1 | - | 1 | 1497 | ||
| BC023882.1 | - | 2 | 559 | 5354698 | ||
| BC025497.1 | - | 2 | 545 | 5320270 | ||
| BC027393.1 | - | 2 | 527 | 4948448 | ||

Haus3
Genomic Map