siRNA Details
| Gene Symbol: Tmem119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146162.2 | NP_666274.1 | 2 | 1627 | ||
| GenBank mRNA | AK078473.1 | - | 2 | 1636 | ||
| AK170712.1 | - | 2 | 1637 | |||
| BC025600.1 | - | 2 | 1632 | 5250899 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem119| Gene Symbol: Tmem119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146162.2 | NP_666274.1 | 2 | 1627 | ||
| GenBank mRNA | AK078473.1 | - | 2 | 1636 | ||
| AK170712.1 | - | 2 | 1637 | |||
| BC025600.1 | - | 2 | 1632 | 5250899 | ||