siRNA Details
Gene Symbol: Tubgcp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146190.2 | NP_666302.2 | 14 | 1827 | ||
GenBank mRNA | AK032031.1 | - | 14 | 1787 | ||
AK044444.1 | - | 14 | 1713 | |||
AK044887.1 | - | 14 | 1811 | |||
AK051326.1 | - | 14 | 1822 | |||
AK086158.1 | - | 14 | 1788 | |||
AK134963.1 | - | 6 | 945 | |||
AK151454.1 | - | 14 | 1785 | |||
AK152033.1 | - | 14 | 1785 | |||
AK153331.1 | - | 14 | 1785 | |||
AK153357.1 | - | 14 | 1785 | |||
AK160939.1 | - | 14 | 1741 | |||
AK161681.1 | - | 15 | 1733 | |||
AK169443.1 | - | 5 | 1171 | |||
AK173291.1 | - | 11 | 1204 | |||
BC027075.1 | - | 6 | 890 | 5355335 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
