siRNA Details
| Gene Symbol: PRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127708.2 | NP_001121180.2 | 4, 5 | 392 | ||
| NM_005807.4 | NP_005798.3 | 5, 6 | 515 | |||
| GenBank mRNA | AK131434.1 | - | 3, 4 | 376 | ||
| AK301343.1 | - | 5, 6 | 510 | |||
| AK301487.1 | - | 3, 4 | 386 | |||
| DA977744.1 | - | 4, 5 | 392 | |||
| U70136.1 | - | 4, 5 | 497 | |||

PRG4
Genomic Map