siRNA Details
| Gene Symbol: Myo3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148413.3 | NP_680779.3 | 1126 | |||
| GenBank mRNA | AY101368.1 | - | 969 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo3a| Gene Symbol: Myo3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148413.3 | NP_680779.3 | 1126 | |||
| GenBank mRNA | AY101368.1 | - | 969 | |||