siRNA Details
| Gene Symbol: GPA33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005814.2 | NP_005805.1 | 7 | 1997 | ||
| XM_017000005.1 | XP_016855494.1 | 8 | 1825 | |||
| GenBank mRNA | BC069705.1 | - | 7 | 1997 | 7262529 | |
| BC069723.1 | - | 7 | 1997 | 7262541 | ||
| BC069745.1 | - | 7 | 1997 | 7262553 | ||
| BC069761.1 | - | 7 | 1997 | 7262565 | ||
| BC069789.1 | - | 7 | 1997 | 7262482 | ||
| BC107164.1 | - | 7 | 1953 | 40034324 | ||
| BC107165.1 | - | 7 | 1955 | 40034331 | ||
| U79725.1 | - | 7 | 1997 | |||

GPA33
Genomic Map