siRNA Details
| Gene Symbol: Zfp472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153063.3 | NP_694703.2 | 4 | 1512 | ||
| GenBank mRNA | AB010338.1 | - | 1 | 101 | ||
| AK142395.1 | - | 4 | 1503 | |||
| AY195875.1 | - | 4 | 1447 | |||
| BC023090.1 | - | 4 | 1447 | |||
| BC027407.1 | - | 4 | 1502 | 4945483 | ||

Zfp472
Genomic Map