siRNA Details
| Gene Symbol: Ehbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252515.1 | NP_001239444.1 | 7 | 945 | ||
| NM_153078.4 | NP_694718.4 | 7 | 945 | |||
| GenBank mRNA | AF424697.1 | - | 6 | 795 | ||
| AK076625.1 | - | 697 | ||||
| AK147102.1 | - | 2 | 268 | |||
| AK173055.1 | - | 7 | 946 | |||

Ehbp1
Genomic Map