siRNA Details
| Gene Symbol: Dnajc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153082.4 | NP_694722.2 | 5 | 685 | ||
| GenBank mRNA | AK049300.1 | - | 5 | 631 | ||
| AY035893.1 | - | 5 | 472 | |||
| BC042643.1 | - | 5 | 600 | 5357595 | ||
| BC051140.1 | - | 5 | 647 | 6595101 | ||
| CB247441.1 | - | 4 | 399 | |||

Dnajc27
Genomic Map