siRNA Details
| Gene Symbol: Hrh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153087.2 | NP_694727.1 | 3 | 1259 | ||
| GenBank mRNA | AF358859.1 | - | 3 | 1259 | ||
| BC111862.1 | - | 2 | 1094 | 40046148 | ||
More Information
Set Membership: |
|

Hrh4
Genomic Map