siRNA Details
| Gene Symbol: St7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253702.1 | NP_001240631.1 | 5 | 1176 | ||
| NM_001253703.1 | NP_001240632.1 | 5 | 1176 | |||
| NM_153091.3 | NP_694731.1 | 5 | 1176 | |||
| GenBank mRNA | AB084518.1 | - | 5 | 590 | ||
| AK033865.1 | - | 5 | 608 | |||
| AK144075.1 | - | 5 | 616 | |||
| AK148099.1 | - | 5 | 878 | |||
| AK172102.1 | - | 5 | 1177 | |||
| AK172218.1 | - | 5 | 1177 | |||
| AK213571.1 | - | 3 | 181 | |||
| AK217761.1 | - | 3 | 181 | |||
| AK219361.1 | - | 3 | 181 | |||
| BC024473.1 | - | 5 | 567 | |||
| BC058781.1 | - | 5 | 601 | 6311899 | ||
| BC107394.2 | - | 5 | 601 | 40058131 | ||
| BC145950.1 | - | 5 | 668 | 40131253 | ||

St7l
Genomic Map