siRNA Details
| Gene Symbol: Fnbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153118.3 | NP_694758.2 | 9, 10 | 1159 | ||
| GenBank mRNA | BC034510.1 | - | 4, 5 | 607 | 4481144 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: AB250296.

Fnbp1l
Genomic Map