siRNA Details
| Gene Symbol: Slc36a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153139.4 | NP_694779.3 | 11 | 2829 | ||
| GenBank mRNA | AY211262.1 | - | 12 | 2004 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc36a1| Gene Symbol: Slc36a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153139.4 | NP_694779.3 | 11 | 2829 | ||
| GenBank mRNA | AY211262.1 | - | 12 | 2004 | ||