siRNA Details
| Gene Symbol: Ggnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153144.2 | NP_694784.2 | 3, 4 | 374 | ||
| GenBank mRNA | AB064543.1 | - | 3, 4 | 374 | ||
| AY633741.1 | - | 3, 4 | 374 | |||
| BC060738.1 | - | 2, 3 | 330 | |||
| BC139772.1 | - | 2, 3 | 280 | 40111779 | ||
| BC145605.1 | - | 2, 3 | 282 | 9054171 | ||
| BC150801.1 | - | 2, 3 | 282 | 9087712 | ||

Ggnbp2
Genomic Map