siRNA Details
Gene Symbol: 2410015M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153152.3 | NP_694792.1 | 4 | 629 | ||
GenBank mRNA | AK075942.1 | - | 4 | 560 | ||
AK075961.1 | - | 4 | 630 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2410015M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153152.3 | NP_694792.1 | 4 | 629 | ||
GenBank mRNA | AK075942.1 | - | 4 | 560 | ||
AK075961.1 | - | 4 | 630 |