siRNA Details
| Gene Symbol: Csrnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153287.3 | NP_695019.1 | 5 | 1109 | ||
| GenBank mRNA | AB091687.1 | - | 4 | 1081 | ||
| AK029893.1 | - | 5 | 1376 | |||
| AK140213.1 | - | 5 | 1122 | |||
| AK171658.1 | - | 4 | 1174 | |||
| BC029720.1 | - | 1 | 151 | |||
| BC050066.1 | - | 5 | 1109 | 30023598 | ||

Csrnp1
Genomic Map